Learn More
Cole-Parmer™ Replacement Impactor for Large Grinding Vial Set (61043-09), Stainless Steel
Catalog No. 6801P
Change view
Click to view available options
Packaging Quantity:
1 Ea.
Catalog No. Packaging Quantity
6801P 1 Ea.
1 options
Catalog No. 6801P Supplier Cole-Parmer™ Supplier No. 6801P
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Replacement impactor for 6801 Large Grinding Vial Set.
  • Made of 440C stainless steel.
  • Withstands liquid nitrogen temperatures.
  • Autoclavable and reusable.
Specifications
Product Type Impactor
Autoclavable Autoclavable
Disposable Reusable
For Use With (Equipment) 6801 Large Grinding Vial Set (61043-09) of SpexSamplePrep Large-Capacity Freezer/Mill
Material Stainless Steel (440 C)
Packaging Quantity 1 Ea.