Learn More
Cole-Parmer™ XRF Spec-Caps™ Cups
These XRF Spec-Caps™ Cups aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques.
Supplier:  Cole-Parmer™ 3617
 View more versions of this product
Catalog No. 0467519
Only null left
Description
Description
Aluminum cups aid in the production of pressed-powder sample discs for XRF.
  • Reinforces sample disks to resist chipping and breaking.
  • Makes sample disks easier to remove from the pellet die.
  • Minimizes contamination during sample handling.
  • Protects the bore of the pellet die from abrasive samples.
  • Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
Specifications
Chemical Identifiers
XRF Spectrometers
Sample Cup
Specifications
XRF Spectrometers
Produces pellets 40 mm x 5 mm
38 mm
9 mm
100 Pk.
Sample Cup
1.5 in.
0.35 in.
Aluminum
Product Suggestions
Product Suggestions