Learn More
Cole-Parmer™ Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer
Catalog No. 2662
Change view
Click to view available options
Holds:
15 x 15 mL Polycarbonate Cryovials
24 x 5 mL Polycarbonate Vials
24 x 5 mL Polyethylene Vials
48 x 1.5 to 2 mL Microcentrifuge or PCR Tubes
6 x 15 mL Short Polycarbonate Vials
6 x 50 mL PC or PE Cryovials
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
2662 24 x 5 mL Polycarbonate Vials
2660 6 x 15 mL Short Polycarbonate Vials
2663 24 x 5 mL Polyethylene Vials
2661 15 x 15 mL Polycarbonate Cryovials
2666 48 x 1.5 to 2 mL Microcentrifuge or PCR Tubes
2664 6 x 50 mL PC or PE Cryovials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 2662 Supplier Cole-Parmer™ Supplier No. 2662
These Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.
  • Safely grind samples in the HG-600 Geno/Grinder™ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
  • Order homogenizer and vials separately.
Specifications
Product Type Holder
For Use With (Application) Ideal for High Throughput Cryogenic Grinding and to Preserve Pesticides, RNA, Proteins and Other Temperature Sensitive Samples
For Use With (Equipment) Spex Geno/Grinder
Holds 24 x 5 mL Polycarbonate Vials
Material Aluminum