Learn More
Cole-Parmer™ Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer
These Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.
Supplier:  Cole-Parmer™ 2660
 View more versions of this product
Catalog No. 26-60
Only null left
Explore available promotions
Description
Description
  • Safely grind samples in the HG-600 Geno/Grinder™ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
  • Order homogenizer and vials separately.
Specifications
Specifications
Holder
Spex Geno/Grinder
Aluminum
Ideal for High Throughput Cryogenic Grinding and to Preserve Pesticides, RNA, Proteins and Other Temperature Sensitive Samples
6 x 15 mL Short Polycarbonate Vials
Product Suggestions
Product Suggestions