missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Grinding Balls for High-Energy Mixer/Mill™ Grinding Vial Sets

Catalog No. 8001B
Change view
Click to view available options
Material:
Hardened Steel
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
8001B Hardened Steel
8007B Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8001B Supplier Cole-Parmer™ Supplier No. 8001B

Crush, grind, and mill various materials with these Grinding Balls for High-Energy Mixer/Mill™ Grinding Vial Sets.

  • Select from 10 different grinding ball materials to suit your application.
  • Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinding vial sets with a wide range of grinding ball options suitable for your application.

Specifications

Product Type Grinding Balls
Composition Fe, Cr, Si, Mn, C
Diameter (English) 0.5 in., 0.25 in.
Diameter (Metric) 1.27 cm, 0.63 cm
For Use With (Application) Ideal for General Purpose Grinding
For Use With (Equipment) Vial Kit of 8000 D and 8000 M Mixer or Mill
Hardness 5.5 to 6
Material Hardened Steel