Learn More
Cole-Parmer™ Vial Set for High-Energy Mixer/Mill™ Grinders
Use this Vial Set with high-energy Mixer/Mill™ grinders to pulverize and mix samples.
Supplier: Cole-Parmer™ SPX8001
Description
- Crush, grind, and mill various materials.
- Optimize performance of your Cole-Parmer™ Freezer/Mill™ grinders with a wide range of vial sets suitable for your application.
Chemical Identifiers
| Grinding Vial Set | |
| 8000 D, 8000 M Mixer/Mills |
Specifications
| Grinding Vial Set | |
| 5.72 cm | |
| 3 in. | |
| Two 1/2 in. and Four 1/4 in. Steel Grinding Balls |
| 2.25 in. | |
| 8000 D, 8000 M Mixer/Mills | |
| 7.62 cm | |
| Hardened Steel |