missing translation for 'onlineSavingsMsg'
Learn More

Spermine, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019831
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019831 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019831 Supplier Merck Emd Millipore Supplier No. 55513100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 71-44-3
Physical Form Neat
Linear Formula NH2(CH2)3 NH(CH2)4 NH(CH2)3 NH2
MDL Number MFCD00008215
Melting Point 28°C to 30°C
Boiling Point 150°C (5 mmHg)
Synonym N,N'-Bis(3-aminopropyl)-1,4-diaminobutane; Gerontine; Musculamine; Neuridine
Quantity 100 mg
Formula Weight 202.34
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less