Learn More
Comecerbrand SpeedyGlove Management and Control Software
Catalog No. 17980816
Change view
Click to view available options
Product Type:
Management Software
Catalog No. Product Type
17980816 Management Software
1 options
Catalog No. 17980816 Supplier Comecerbrand Supplier No. NL0154010001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
SpeedyGlove Management and Control software. OPTION.
  • Optional client-server, web-based solution that provides flexibility and traceability when operating multiple Speedy Glove units in high-volume pharmaceutical or production environments.
  • The software is easy to use, and once all required data—such as glove port adapters, glove ports, glove types, recipes, and configurations—has been entered, it significantly improves the efficiency of glove testing.
  • When used with the associated RFID tags (P/N: NL0254020007), the software enables the Speedy Glove to automatically identify the correct recipe for each glove port and prompt the operator to start the test.
  • After the test is completed (typically within 7 to 10 minutes), the report is automatically saved to the server (21 CFR Part 11 compliant) and linked to the corresponding glove port, adapter, and Speedy Glove unit for complete traceability and informed glove replacement.
Specifications
Product Type Management Software