Learn More
United Scientific™ Spectrum Tubes
The classic demonstration of spectral lines.
Supplier:  United Scientific™ SPTNE01
 View more versions of this product
Catalog No. S06996
Only null left
Description
Description
Acquaint students with the spectra of common gases in their pure form while demonstrating concept of wavelength.
  • 25 cm (10 in.) long
  • Capillary portion about 4 in. (10 cm) long
Specifications
Chemical Identifiers
5 to 12
Spectrometer
10 in.
4 in.
Emits characteristic spectrum when energized by spectroscope
Sealed electrode, metal endcap, loop and activity guide
25.4 cm
10.16 cm
Specifications
5 to 12
Spectrometer
10 in.
4 in.
Neon Spectrum Tube
Emits characteristic spectrum when energized by spectroscope
Sealed electrode, metal endcap, loop and activity guide
25.4 cm
10.16 cm
Product Suggestions
Product Suggestions