missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing

Catalog No. 0880127
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
15 mm
16 mm
20 mm
24 mm
MWCO:
0.1 to 0.5 kDa
0.5 kDa
1 kDa
10 kDa
100 kDa
25 kDa
5 kDa
50 kDa
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) MWCO
0880127 15 mm 25 kDa
0867125 20 mm 0.5 kDa
0867127 20 mm 1 kDa
0867124 24 mm 0.5 kDa
0867126 15 mm 1 kDa
0867130 15 mm 5 kDa
0867131 20 mm 5 kDa
0867132 15 mm 10 kDa
0867133 20 mm 10 kDa
0867134 6.4 mm 50 kDa
0867135 10 mm 50 kDa
0867136 15 mm 50 kDa
0867137 20 mm 50 kDa
0867138 6.4 mm 100 kDa
0867139 10 mm 100 kDa
0867140 15 mm 100 kDa
0867141 20 mm 100 kDa
0875010B 6.4 mm 25 kDa
087502D 10 mm 0.1 to 0.5 kDa
087502B 6.4 mm 0.5 kDa
0875030B 10 mm 25 kDa
087503D 16 mm 1 kDa
087506B 6.4 mm 5 kDa
087506D 10 mm 5 kDa
087508B 6.4 mm 10 kDa
087508D 10 mm 10 kDa
087503B 6.4 mm 1 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
Catalog No. 0880127 Supplier Spectrum™ Supplier No. 131345

Selected MWCOs from 100 to 100,000 Daltons

  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution

Order Info

Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.

Specifications

Product Type Spectra/Por Dialysis Membrane Tubing
MWCO 25 kDa
Material Cellulose Ester
Diameter (Metric) 15 mm
Diameter (English) 0.59 in.
Length (English) 33 ft.
Length (Metric) 10 m
Formulation Supplied in 0.1% sodium azide solution
Temperature Range (English) 39.2°F to 98.6°F
Temperature Range (Metric) 4°C to 37°C
Quantity Each
For Use With (Application) Ideal for desalting, purification, and molecular separation
Show More Show Less