missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ Biotech Cellulose Ester (CE) Dialysis Membrane Tubing

Selected MWCOs from 100 to 100,000 Daltons

Supplier:  Spectrum™ 131336

 View more versions of this product

Catalog No. 08-750-10B

Only null left
Explore available promotions

Description

Description

Selected MWCOs from 100 to 100,000 Daltons

  • CE tubing membranes are ideal for desalting, purification, and molecular separation
  • Supplied in 10m (33 ft.) lengths in two rolls of 5m each, in 0.1% sodium azide solution

Order Info

Note:Do not tie—must be used with Spectra/Por nylon closures (08-671-52, -53, -55). Strong polar solvents such as acetone, MEK, or dioxane will irreparably damage membranes. Contact Spectrum Technical Service, 1-800-634-3300, for more information.

Specifications

Specifications

Spectra/Por Dialysis Membrane Tubing
Cellulose Ester
0.25 in.
10 m
39.2°F to 98.6°F
Each
25 kDa
6.4 mm
33 ft.
Supplied in 0.1% sodium azide solution
4°C to 37°C
Ideal for desalting, purification, and molecular separation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions