Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Spectra/Por™ 7 Membrane Tubing (Sulfur and Heavy Metal Free):1 1000 to 50,000 Dalton MWCO

Catalog No. 086809E
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Diameter (Metric):
7.5 mm
11.5 mm
15 mm
17.8 mm
20.4 mm
21.6 mm
24 mm
25.5 mm
29 mm
32 mm
34.4 mm
45 mm
Quantity:
Each
MWCO:
1 kDa
10 kDa
15 kDa
2 kDa
25 kDa
3.5 kDa
50 kDa
8 kDa
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity MWCO
086809E 21.6 mm Each 50 kDa
086802E 24 mm Each 1 kDa
086802C 11.5 mm Each 1 kDa
086802G 45 mm Each 1 kDa
086803E 24 mm Each 2 kDa
086803G 29 mm Each 2 kDa
086803C 11.5 mm Each 2 kDa
086804G 34.4 mm Each 3.5 kDa
086804E 29 mm Each 3.5 kDa
086805A 7.5 mm Each 8 kDa
086805C 15 mm Each 8 kDa
086805J 32 mm Each 8 kDa
086805E 20.4 mm Each 8 kDa
086805G 25.5 mm Each 8 kDa
086806A 7.5 mm Each 10 kDa
086806E 20.4 mm Each 10 kDa
086806G 29 mm Each 10 kDa
086807A 7.5 mm Each 15 kDa
086807C 15 mm Each 15 kDa
086808A 7.5 mm Each 25 kDa
086808C 17.8 mm Each 25 kDa
086807G 29 mm Each 15 kDa
086808E 21.6 mm Each 25 kDa
086807E 20.4 mm Each 15 kDa
086809A 7.5 mm Each 50 kDa
086809C 17.8 mm Each 50 kDa
086806C 15 mm Each 10 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 missing translation for 'options'
Catalog No. 086809E missing translation for 'mfr' Spectrum™ missing translation for 'supplierNo' 132130

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por 7 Membrane Tubing
MWCO 50 kDa
Material Regenerated Cellulose
Diameter (Metric) 21.6 mm
Diameter (English) 0.85 in.
Length (English) 16.4 ft.
Length (Metric) 5 m
Color Transparent
Formulation Packaged wet in 0.05% sodium azide
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less