missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por™ 3 RC Dialysis Membrane Tubing 3500 Dalton MWCO
Description
Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.
- Transparent and flexible
- Low molecular weight compounds in aqueous solutions diffuse through membrane
- Higher molecular weights, such as proteins and bacteria, are retained by membrane
- Glycerol preserves membrane and must be removed before use
- Flat sheets and discs available for use with dialysis apparatus
Do not tie knots in tubing; use closures
Specifications
Specifications
| Product Type | Spectra/Por 3 Dialysis Tubing |
| MWCO | 3.5 kDa |
| Material | Regenerated Cellulose |
| Diameter (Metric) | 45 mm |
| Diameter (English) | 1.77 in. |
| Length (English) | 49.2 ft. |
| Length (Metric) | 15 m |
| Formulation | Contain glycerin as a humectant |
| Temperature Range (English) | 39.2°F to 249.8°F |
| Temperature Range (Metric) | 4°C to 121°C |
| Show More |