missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 2 RC Dialysis Membrane Tubing 12,000 to 14,000 Dalton MWCO

Catalog No. p-86931
Change view
Click to view available options
Diameter (Metric):
6.4 mm
16 mm
45 mm
67 mm
76 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
086703BB 16 mm
086703C 76 mm
086703B 67 mm
086703AA 6.4 mm
086703A 45 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 086703BB Supplier Spectrum™ Supplier No. 132678
Only null left

Ideal for molecular separation, protein binding studies, electrolyte removal, desalting

Supplied as flat-width tubing or flat sheets that can be folded to fit various funnel-type dialysis apparatus. Pure regenerated natural cellulose membranes with 1000 to 50,000 molecular weight cutoffs.

  • Transparent and flexible
  • Low molecular weight compounds in aqueous solutions diffuse through membrane
  • Higher molecular weights, such as proteins and bacteria, are retained by membrane
  • Glycerol preserves membrane and must be removed before use
  • Flat sheets and discs available for use with dialysis apparatus

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por 2 Dialysis Tubing
MWCO 12 to 14 kDa
Material Regenerated Cellulose
Diameter (Metric) 16 mm
Diameter (English) 0.62 in.
Length (English) 49.2 ft.
Length (Metric) 15 m
Formulation Contain glycerin as a humectant
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less