missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por Biotech-Grade RC Dialysis Membrane Tubing

Catalog No. 08607068
Change view
Click to view available options
Diameter (Metric):
6.4 mm
10 mm
MWCO:
20 kDa
3.5 to 5 kDa
8 to 10 kDa
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) MWCO
08607068 10 mm 20 kDa
08607063 6.4 mm 3.5 to 5 kDa
08607064 10 mm 3.5 to 5 kDa
08607065 6.4 mm 8 to 10 kDa
08607066 10 mm 8 to 10 kDa
08607067 6.4 mm 20 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 08607068 Supplier Spectrum™ Supplier No. 133342

A premium dialysis membrane that provides superior molecular selectivity than Standard RC and superior tolerance than Biotech CE

  • Crafted from a regeneration process that yields superior temperature tolerances and chemical compatibilities with the same high purity and selectivity as Biotech CE
  • Membrane can tolerate concentrated-weak acids and bases, dilute-strong acids and bases, most alcohols and some mild or dilute organic solvents
  • Rigidly controlled porosity for better selectivity and separations
  • Ultrapure, does not interfere with molecular activity and function
  • 3 MWCOs and 2 tubing flat-widths
  • Packaged dry with glycerin (soak 15 min. in water)
  • Use with pH 2 to 12 and temp 4 to 60°C

Specifications

Product Type Spectra/Por Dialysis Tubing
MWCO 20 kDa
Material Regenerated Cellulose
Diameter (Metric) 10 mm
Diameter (English) 0.39 in.
Length (English) 16.4 ft.
Length (Metric) 5 m
Formulation Packaged dry with glycerin (soak 15 min. in water)
Temperature Range (English) 39.2°F to 140°F
Temperature Range (Metric) 4°C to 60°C
Quantity Each
For Use With (Application) Frequent dialysis applications, desalting, purification, sample preparation
Show More Show Less