missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 6 Pre-wetted Standard RC Dialysis Tubing, 1-50 kD MWCO

Catalog No. p-86984
Change view
Click to view available options
Diameter (Metric):
7.5 mm
11.5 mm
15 mm
17.8 mm
20.4 mm
21.6 mm
24 mm
29 mm
31.8 mm
34.4 mm
MWCO:
1 kDa
10 kDa
15 kDa
2 kDa
25 kDa
3.5 kDa
50 kDa
8 kDa
25 product options available for selection
Product selection table with 25 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) MWCO
0867012B 11.5 mm 1 kDa
0867012C 24 mm 1 kDa
0867012D 29 mm 1 kDa
0867013B 11.5 mm 2 kDa
0867013C 24 mm 2 kDa
0867014A 11.5 mm 3.5 kDa
0867013D 29 mm 2 kDa
0867014C 34.4 mm 3.5 kDa
0867015A 15 mm 8 kDa
0867015B 20.4 mm 8 kDa
0867015D 31.8 mm 8 kDa
0867016A 7.5 mm 10 kDa
0867016B 15 mm 10 kDa
0867016C 20.4 mm 10 kDa
0867016D 29 mm 10 kDa
0867017A 7.5 mm 15 kDa
0867017B 15 mm 15 kDa
0867017C 20.4 mm 15 kDa
0867018A 7.5 mm 25 kDa
0867018B 17.8 mm 25 kDa
0867019C 21.6 mm 50 kDa
0867019B 17.8 mm 50 kDa
0867019A 7.5 mm 50 kDa
0867018C 21.6 mm 25 kDa
0867014B 29 mm 3.5 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 25 options available.
25 options
Catalog No. 0867012B Supplier Spectrum™ Supplier No. 132636
Only null left

Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane

This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.

  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por 6 Dialysis Tubing
MWCO 1 kDa
Material Regenerated Cellulose
Diameter (Metric) 11.5 mm
Diameter (English) 0.45 in.
Length (English) 33 ft.
Length (Metric) 10 m
Formulation Packaged wet in 0.05% sodium azide
Temperature Range (English) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantity Each
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less