missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ 1-4 Standard RC Dialysis Membrane in Flat Sheets

Catalog No. p-87169
Change view
Click to view available options
MWCO:
12 to 14 kDa
3.5 kDa
6 to 8 kDa
Width (English):
4.2 in.
7.8 in.
9.4 in.
Length (English):
4.2 in.
7.8 in.
9.4 in.
Membrane:
Spectra/Por 1
Spectra/Por 2
Spectra/Por 3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Width (English) Length (English) Membrane
086704 12 to 14 kDa 7.8 in. 7.8 in. Spectra/Por 2
0867120 3.5 kDa 4.2 in. 4.2 in. Spectra/Por 3
0868254A 6 to 8 kDa 9.4 in. 9.4 in. Spectra/Por 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 086704 Supplier Spectrum™ Supplier No. 132686
Only null left

Clear, flexible and sturdy Standard RC (regenerated cellulose) in single layer square sheets that can be used with a variety of dialysis equipment or can be cut into specific dimensions for attaching to support frames for customized diagnostic assays, equilibrium studies, desalting, protein purification, etc.

  • Flat-Sheets deal for use with dialysis equipment or cutting to size
  • Available in 3.5 kD, 6-8 kD and 12-14 kD MWCO's
  • Good chemical compatibility with dilute strong acids & bases, conc. weak acids and bases, many alcohols & some organics.
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • 25 sheets/pkg

Do not tie knots in tubing; use closures

Specifications

Product Type Spectra/Por 2 Dialysis Membrane Flat Sheets
MWCO 12 to 14 kDa
Membrane Spectra/Por 2
Length (English) 7.8 in.
Length (Metric) 200 mm
Width (English) 7.8 in.
Width (Metric) 200 mm
Dimensions (L x W) 7.8 x 7.8 in. (200 x 200 mm)
Color Clear
Temperature Range (English) 39°F to 250°F
Temperature Range (Metric) 4°C to 121°C
Quantity 25 Sheets
For Use With (Application) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting
Show More Show Less