missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device

Catalog No. p-4412009
Change view
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
20 kDa
3.5 to 5 kDa
50 kDa
8 to 10 kDa
Closure Color:
Black
Blue
Green
Orange
Red
White
Yellow
Height (Metric):
44 mm
63 mm
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Closure Color Height (Metric)
08607072 0.1 to 0.5 kDa Green 44 mm
08607073 0.5 to 1 kDa Orange 44 mm
08607074 3.5 to 5 kDa Black 44 mm
08607075 8 to 10 kDa Yellow 44 mm
08607076 20 kDa Red 44 mm
08607077 50 kDa White 44 mm
08607079 0.1 to 0.5 kDa Green 63 mm
08607078 100 kDa Blue 44 mm
08607080 0.5 to 1 kDa Orange 63 mm
08607081 3.5 to 5 kDa Black 63 mm
08607082 8 to 10 kDa Yellow 63 mm
08607083 20 kDa Red 63 mm
08607084 50 kDa White 63 mm
08607085 100 kDa Blue 63 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Catalog No. 08607072 Supplier Spectrum™ Supplier No. F235049
Only null left

Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever

Ready-to-Use and disposable, the Float-A-Lyzer G2 incorporates Biotech Grade Cellulose Ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use & provides ultimate convenience. Available in volume sizes: 1, 5, and 10 mL. The resealable screw cap is color-coded for 9 available MWCO's and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5 and 10mL sizes). The flotation ring (included) provides buoyancy to maximize dialysis efficiency.

  • Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
  • Leak-proof Luer-Lok™ cap provides easy access with the included syringe for easy loading and sample retrieval
  • Units can also interlock for the simultaneous dialysis of multiple samples
  • Ready-to-use and complete
  • Easy syringe loading (no needle)
  • Total sample recovery
  • Ultrapure Biotech CE (no cleaning)
  • Largest range of MWCOs
  • Maximizes SA & minimizes hold-up volume

Specifications

Product Type Micro Float-A-Lyzer Dialysis Device
MWCO 0.1 to 0.5 kDa
Membrane Cellulose Ester
Material Polycarbonate
Diameter (English) Membrane 0.25 in.
Diameter (Metric) Membrane 6.4 mm
Height (English) 1.7 in.
Height (Metric) 44 mm
Width (English) 1.77 in.
Width (Metric) 45 mm
Capacity (Metric) 100 to 200 μL
Closure Color Green
Closure Material Polypropylene
Closure Type Luer lok
Kit Contents 1mL syringe
Quantity Each
For Use With (Application) Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
Show More Show Less