missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Spectra/Por™ RC Dialysis Discs
$907.32 - $907.32
Specifications
| Product Type | RC Dialysis Disc |
|---|---|
| Material | Regenerated Cellulose |
| Quantity | 50 Pack |
| For Use With (Application) | For laboratory dialysis functions such as desalting, buffer exchange or molecular separation |
Description
- Material: Regenerated Cellulose
- Hydrophilic
- Symmetric porosity
- Carries no fixed charge
- Does not adsorb most solutes
- pH limits: 2 to 12
- Packaged dry with glycerol (must be rinsed out with water)
For laboratory dialysis functions such as desalting, buffer exchange or molecular separation
Specifications
| RC Dialysis Disc | |
| Regenerated Cellulose | |
| 50 Pack | |
| For laboratory dialysis functions such as desalting, buffer exchange or molecular separation |