missing translation for 'onlineSavingsMsg'
Learn More

Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices

Catalog No. 08607007
Change view
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
1000 kDa
20 kDa
3.5 to 5 kDa
300 kDa
50 kDa
8 to 10 kDa
Color Code:
Amber
Black
Blue
Green
Orange
Pink
Red
Violet
Yellow
Length (Metric):
50 mm
100 mm
160 mm
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO Color Code Length (Metric)
08607007 20 kDa Red 100 mm
08607012 100 kDa Blue 50 mm
08607023 3.5 to 5 kDa Black 50 mm
08607010 50 kDa Violet 160 mm
08607006 100 kDa Blue 100 mm
08607027 300 kDa Amber 50 mm
08607022 0.1 to 0.5 kDa Green 50 mm
08607021 8 to 10 kDa Yellow 50 mm
08607016 0.1 to 0.5 kDa Green 160 mm
08607031 1000 kDa Pink 100 mm
08607005 50 kDa Violet 50 mm
08607015 8 to 10 kDa Yellow 160 mm
08607032 1000 kDa Pink 160 mm
08607001 0.1 to 0.5 kDa Green 100 mm
08607026 0.5 to 1 kDa Orange 160 mm
08607028 300 kDa Amber 100 mm
08607008 50 kDa Violet 100 mm
08607025 0.5 to 1 kDa Orange 100 mm
08607030 1000 kDa Pink 50 mm
08607029 300 kDa Amber 160 mm
08607024 0.5 to 1 kDa Orange 50 mm
08607020 3.5 to 5 kDa Black 100 mm
08607009 20 kDa Red 160 mm
08607000 8 to 10 kDa Yellow 100 mm
08607017 3.5 to 5 kDa Black 160 mm
08607011 100 kDa Blue 160 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. 08607007 Supplier Spectrum™ Supplier No. G235057

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)

Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.

  • Superior handling and leak protection
  • Resealable screw cap and pipet for easy loading and total recovery
  • Highest membrane purity, no heavy metals and sulfides
  • Minimizes volume specific dilution
  • 9 MWCOs ranging from 100 to 1,000,000 Daltons

Specifications

Product Type Float-A-Lyzer G2 Dialysis Device
MWCO 20 kDa
Membrane Cellulose Ester
Material Polypropylene
Diameter (English) Membrane 0.39 in.
Diameter (English)Top 0.9 in.
Diameter (Metric) Membrane 10 mm
Diameter (Metric)Top 23 mm
Length (English) 3.9 in.
Length (Metric) 100 mm
Closure Material Polypropylene
Closure Type Screw Cap
Color Code Red
Kit Contents Disposable pipette for 5 and 10mL, flotation ring
Quantity 12 Pack
For Use With (Application) Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions
Show More Show Less