missing translation for 'onlineSavingsMsg'
Learn More
Learn More
- Home
- Products
- Protein Analysis Reagents
- Dialysis and Desalting
- General Dialysis Supplies
- Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Catalog No. 08607005
Change view
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
1000 kDa
20 kDa
3.5 to 5 kDa
300 kDa
50 kDa
8 to 10 kDa
Color Code:
Amber
Black
Blue
Green
Orange
Pink
Red
Violet
Yellow
Length (Metric):
50 mm
100 mm
160 mm
26 product options available for selection
| Catalog No. | MWCO | Color Code | Length (Metric) |
|---|---|---|---|
| 08607005 | 50 kDa | Violet | 50 mm |
| 08607012 | 100 kDa | Blue | 50 mm |
| 08607023 | 3.5 to 5 kDa | Black | 50 mm |
| 08607010 | 50 kDa | Violet | 160 mm |
| 08607006 | 100 kDa | Blue | 100 mm |
| 08607027 | 300 kDa | Amber | 50 mm |
| 08607022 | 0.1 to 0.5 kDa | Green | 50 mm |
| 08607021 | 8 to 10 kDa | Yellow | 50 mm |
| 08607016 | 0.1 to 0.5 kDa | Green | 160 mm |
| 08607031 | 1000 kDa | Pink | 100 mm |
| 08607015 | 8 to 10 kDa | Yellow | 160 mm |
| 08607032 | 1000 kDa | Pink | 160 mm |
| 08607001 | 0.1 to 0.5 kDa | Green | 100 mm |
| 08607026 | 0.5 to 1 kDa | Orange | 160 mm |
| 08607028 | 300 kDa | Amber | 100 mm |
| 08607008 | 50 kDa | Violet | 100 mm |
| 08607025 | 0.5 to 1 kDa | Orange | 100 mm |
| 08607030 | 1000 kDa | Pink | 50 mm |
| 08607029 | 300 kDa | Amber | 160 mm |
| 08607024 | 0.5 to 1 kDa | Orange | 50 mm |
| 08607007 | 20 kDa | Red | 100 mm |
| 08607020 | 3.5 to 5 kDa | Black | 100 mm |
| 08607009 | 20 kDa | Red | 160 mm |
| 08607000 | 8 to 10 kDa | Yellow | 100 mm |
| 08607017 | 3.5 to 5 kDa | Black | 160 mm |
| 08607011 | 100 kDa | Blue | 160 mm |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
Catalog No.
08607005
Supplier
Spectrum™
Supplier No.
G235034
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.
- Superior handling and leak protection
- Resealable screw cap and pipet for easy loading and total recovery
- Highest membrane purity, no heavy metals and sulfides
- Minimizes volume specific dilution
- 9 MWCOs ranging from 100 to 1,000,000 Daltons
Specifications
Specifications
| Product Type | Float-A-Lyzer G2 Dialysis Device |
| MWCO | 50 kDa |
| Membrane | Cellulose Ester |
| Diameter (English) Membrane | 0.39 in. |
| Diameter (English)Top | 0.9 in. |
| Diameter (Metric) Membrane | 10 mm |
| Diameter (Metric)Top | 23 mm |
| Length (English) | 1.9 in. |
| Length (Metric) | 50 mm |
| Closure Material | Polypropylene |
| Closure Type | Screw Cap |
| Color Code | Violet |
| Kit Contents | Disposable pipette for 5 and 10mL, flotation ring |
| Quantity | 12 Pack |
| For Use With (Application) | Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions |
| Show More Show Less |