Learn More
Spectrum™ Spectra/Por™ Float-A-Lyzer™ G2 Dialysis Devices
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Supplier: Spectrum™ G235049
Description
Ready-to-use and disposable, Float-A-Lyzer G2 incorporates biotech-grade cellulose ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use and provides ultimate convenience for small volume dialysis (1 to 10mL)
Available in volume sizes: 1mL, 5mL and 10 mL. Resealable screw cap is color-coded for 9 available MWCOs and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5mL and 10mL sizes). Flotation ring (included) provides buoyancy to maximize dialysis efficiency.
- Superior handling and leak protection
- Resealable screw cap and pipet for easy loading and total recovery
- Highest membrane purity, no heavy metals and sulfides
- Minimizes volume specific dilution
- 9 MWCOs ranging from 100 to 1,000,000 Daltons
Specifications
| Float-A-Lyzer G2 Dialysis Device | |
| Cellulose Ester | |
| 0.9 in. | |
| 23 mm | |
| 100 mm | |
| Screw Cap | |
| Disposable pipette for 5 and 10mL, flotation ring | |
| Ready to use laboratory dialysis devices, highest membrane purity, superior handling and leak prevention, 95 to 98% sample recovery, perfect for dialyzing electro-eluted proteins or chromatography fractions |
| 0.1 to 0.5 kDa | |
| 0.39 in. | |
| 10 mm | |
| 3.9 in. | |
| Polypropylene | |
| Green | |
| 12 Pack |