missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectinomycin Sulfate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDE 34:2009 (AClass Cert AR-1470).
Specifications
Specifications
| CAS | 64058-48-6 |
| MDL Number | MFCD04041129 |
| Synonym | Spectinomycin |
| Type | Certified Reference Material |
| Quantity | 1 g |
| Grade | Certified Reference Material |
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to pharmacopeia primary standards. These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. This product was designed, produced and verified for accuracy and stability in accordance with ISO/IEC 17025:2005 (AClass Cert AT-1467), ISO GUIDE 34:2009 (AClass Cert AR-1470).