missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin sulfate, 93%

Catalog No. AAJ6603406
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ6603406 5 g Plastic bottle
AAJ6603403 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6603406 Supplier Thermo Scientific Chemicals Supplier No. J6603406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23312-56-3 | C14H26N2O11S | 430.43 g/mol

Targets Gram-negative bacteria

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23312-56-3
Molecular Formula C14H26N2O11S
Molecular Weight (g/mol) 430.43
MDL Number MFCD00270185
InChI Key XGBFWQUQYQIFLB-MTTMTQIXSA-N
Synonym spectinomycin sulfate, spectinomycin sulphate, unii-bz0h4tlf9x, actinospectacin, trobicin, bz0h4tlf9x, togamycin sulfate 1:1, spectinomycin sulfate hydrate, spectinomicina, spectogard
PubChem CID 64771
IUPAC Name (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one; sulfuric acid
SMILES OS(O)(=O)=O.CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O

Specifications

Boiling Point 583°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 5 g
Solubility Information Soluble in DMSO or water
Formula Weight 430.43
Percent Purity 93%
Physical Form Powder
Chemical Name or Material Spectinomycin sulfate
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
EINECSNumber 245-578-7
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only