missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectinomycin Dihydrochloride Pentahydrate 98.0+%, TCI America™
$144.30 - $527.82
Chemical Identifiers
| CAS | 22189-32-8 |
|---|---|
| Molecular Formula | C14H36Cl2N2O12 |
| Molecular Weight (g/mol) | 495.34 |
| MDL Number | MFCD00150886 |
| InChI Key | DCHJOVNPPSBWHK-UXXUFHFZSA-N |
| PubChem CID | 67108398 |
| ChEBI | CHEBI:9217 |
| IUPAC Name | dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride |
| SMILES | [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
Chemical Identifiers
| 22189-32-8 | |
| 495.34 | |
| DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
| CHEBI:9217 | |
| [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
| C14H36Cl2N2O12 | |
| MFCD00150886 | |
| 67108398 | |
| dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride |
Specifications
| Spectinomycin Dihydrochloride Pentahydrate | |
| 22189-32-8 | |
| White | |
| MFCD00150886 | |
| [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O | |
| 495.34 | |
| CHEBI:9217 | |
| ≥98.0% (N,T) |
| Crystalline Powder | |
| 207°C | |
| C14H36Cl2N2O12 | |
| DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
| dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride | |
| 67108398 | |
| 405.27 | |
| Spectinomycin Dihydrochloride Pentahydrate |
Safety and Handling
RTECSNumber : WG7400000
TSCA : No
Recommended Storage : Refrigerator