missing translation for 'onlineSavingsMsg'
Learn More

Spectinomycin Dihydrochloride Pentahydrate 98.0+%, TCI America™

Supplier:  TCI America S058425G

Quantity 25 g

 View more versions of this product

Catalog No. S058425G


Only null left

Description

Description

Specifications

Chemical Identifiers

22189-32-8
495.34
DCHJOVNPPSBWHK-UXXUFHFZSA-N
CHEBI:9217
[H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
C14H36Cl2N2O12
MFCD00150886
67108398
dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride

Specifications

Spectinomycin Dihydrochloride Pentahydrate
Crystalline Powder
207°C
25 g
MFCD00150886
[H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
495.34
CHEBI:9217
≥98.0% (N,T)
Spectinomycin Dihydrochloride Pentahydrate
22189-32-8
White
C14H36Cl2N2O12
DCHJOVNPPSBWHK-UXXUFHFZSA-N
dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one pentahydrate dichloride
67108398
405.27
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : WG7400000

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions