Learn More
Spectinomycin dihydrochloride pentahydrate, Cell Culture Grade
Spectinomycin dihydrochloride pentahydrate, CAS # 22189-32-8, is the hydrochloride salt form of spectinomycin, an aminocyclitol compound showing antibacterial and antimicrobial activities. | CAS: 22189-32-8 | C14H36Cl2N2O12 | 495.34 g/mol
Supplier: Thermo Scientific Chemicals J6182006
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
General Description
- Spectinomycin dihydrochloride pentahydrate is derived from Streptomyces spectabilis and presents antibacterial and antimicrobial activities
- Spectinomycin hydrochloride is a compound that inhibits the 30S ribosomal subunit of bacteria and interrupts protein synthesis and peptide elongation, leading to bacterial cell death
Application
- Spectinomycin dihydrochloride pentahydrate is a compound that is active against gram-negative bacteria
- In the laboratory, this compound is used in cell culture applications
- It prevents contamination and can be used to select for resistant bacteria and plant cells containing the marker gene Spcr
- It has been used to study respiratory tract infections caused by Pasteurella multocida and Mannheimia haemolytica
- In genetic approaches, this compound can be used to amplify the plasmid copy number
Specifications
| Spectinomycin Dihydrochloride Pentahydrate | |
| Powder | |
| White | |
| C14H36Cl2N2O12 | |
| Spectinomycin dihydrochloride | |
| DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
| dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,â¸]tetradecan-7-one pentahydrate dichloride | |
| 67108398 | |
| 495.35 (405.28 Anhydrous) |
| Spectinomycin dihydrochloride pentahydrate | |
| 22189-32-8 | |
| 5 g | |
| MFCD00150886 | |
| Soluble in water;Soluble in water,methanol,dimethyl sulfoxide,ethanol and dimethyl formamide. | |
| [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O | |
| 495.34 | |
| CHEBI:9217 | |
| Cell Culture |
Chemical Identifiers
| 22189-32-8 | |
| 495.34 | |
| DCHJOVNPPSBWHK-UXXUFHFZSA-N | |
| 67108398 | |
| dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,â¸]tetradecan-7-one pentahydrate dichloride |
| C14H36Cl2N2O12 | |
| MFCD00150886 | |
| Spectinomycin dihydrochloride | |
| CHEBI:9217 | |
| [H+].[H+].O.O.O.O.O.[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 244-554-3
RTECSNumber : WG7400000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only