missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Specimen Tape
GREENER_CHOICE

For labeling and identifying specimens.

$12.90

Specifications

Color White
Format Tape
For Use With (Application) Labeling and identifying specimens
Quantity 1 Ea.
Width (English) 1 in.
View More Specs

Products 2
Catalog Number Display Text Price Quantity & Availability  
Catalog Number Display Text Price Quantity & Availability  
15929
GREENER_CHOICE
View Documents Promotion Details
Name, Room No., Hospital No., Date
Each for $12.90
Only null left
 
15930
View Documents Promotion Details
Name, Room No., Date
Each for $12.90
Only null left
 
Description

Description

  • White tape printed with headings for Name, Room No., and Date, followed by space for notations
  • Also available with additional printed heading for Hospital No.

Order Info

Label size, 2.25 x 1 in. W (5.7 x 2.5cm). Sold without dispenser (fits Fisherbrand dispenser 15-942).

Specifications

Specifications

White
Labeling and identifying specimens
1 in.
Tape
1 Ea.
Videos
SDS
Documents

Documents