missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Specimen Tape

Catalog No. 15930
Change view
Click to view available options
Display Text:
Name, Room No., Date
Name, Room No., Hospital No., Date
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Display Text
15930 Name, Room No., Date
15929 Name, Room No., Hospital No., Date
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15930

For labeling and identifying specimens.

  • White tape printed with headings for Name, Room No., and Date, followed by space for notations
  • Also available with additional printed heading for Hospital No.

Order Info

Label size, 2.25 x 1 in. W (5.7 x 2.5cm). Sold without dispenser (fits Fisherbrand dispenser 15-942).

Specifications

Color White
Format Tape
Display Text Name, Room No., Date
For Use With (Application) Labeling and identifying specimens
Quantity 1 Ea.
Width (English) 1 in.