Learn More
GSC Go Science Crazy Specimen Jars
Catalog No. S24595
Change view
Click to view available options
Capacity (English):
8 oz.
Closure Size:
70-400 mm
Catalog No. Capacity (English) Closure Size
S24595 8 oz. 70-400 mm
1 options
Catalog No. S24595 Supplier GSC Go Science Crazy Supplier No. EPD4104
Only null left
For storage and dispensing specimens. GSC International Specimen Jars are made of glass for its optical qualities of light transmission and chemical resistance making them perfect for lab use.
  • Made of clear flint glass
  • Includes plastic cap
  • Wide mouth
Specifications
Capacity (English) 8 oz.
Includes Cap
Material Flint Glass
Liner Material Foam
Mouth Wide
Product Type Specimen Jar
Closure Material Plastic
Closure Size 70-400 mm
Closure Type Screw Cap