Learn More
Mopec™ Specimen Chucks for CRYOSTAR Cryostats, 40 mm
Specimen Chucks are available in multiple sizes and colors. Some chucks are also available in waffle pattern.
Supplier: Mopec™ SP124WWH
Description
- Round, aluminum specimen chucks
Chemical Identifiers
| CryoStar NX50 and NX70 | |
| Aluminum |
Specifications
| Waffle Chuck | |
| CryoStar NX50 and NX70 | |
| Aluminum | |
| White |