missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Specialty Glass Column: Glass Coarse Frit

Described in US EPA Protocol - Fill columns with appropriate adsorbents when cleaning or drying environmental samples for pesticide or priority pollutant analyses

Supplier:  MilliporeSigma™ Supelco™ 64755

 View more versions of this product

Catalog No. 111010687


Only null left

Description

Description

Specifications

Chemical Identifiers

LPLC
Glass (Frit), PTFE (Stopcock)
1
Frit

Specifications

Specialty Glass Column
LPLC
1
22 mm
25 mm
Glass (Frit), PTFE (Stopcock)
Overall Length: 515 mm
Frit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions