missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SPATA16 Polyclonal Antibody

Description
SPATA16 Polyclonal Antibody for Western Blot, IP, ChIP
This gene encodes a testis-specific protein that belongs to the tetratricopeptide repeat-like superfamily. The encoded protein localizes to the Golgi apparatus and may play a role in spermatogenesis.
Specifications
Specifications
| Antigen | SPATA16 |
| Applications | ChIP Assay, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | SPATA16 |
| Gene Accession No. | Q8C636, Q9BXB7 |
| Gene Alias | 4921511F01Rik; 4930503K02Rik; CDC68; FACT; FACTP140; FLJ10857; FLJ14010; NYD-SP12; RGD1563726; Spata16; spermatogenesis associated 16; spermatogenesis-associated protein 16; spermatogenesis-related protein; SPGF6; SUPT16H; testicular tissue protein Li 185; testis development protein NYD-SP12; testis-specific Golgi protein |
| Gene Symbols | SPATA16 |
| Show More |