Learn More
Eisco™ Spare Extractors for Soxhlet Apparatus
Sockets and cones feature ground glass. A variety of sizes available
Supplier: Eisco™ CH0888J
Description
Specifications
| 1 | |
| 200 mL | |
| Distillation | |
| 9 cm |
| Socket size 50/42, Cone size 24/29 | |
| Borosilicate Glass | |
| 37 cm |
1 year limited manufacturer warranty