missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-DTAF (5-(4,6-Dichlorotriazinyl) Aminofluorescein), single isomer

Catalog No. D16
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D16 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D16 Supplier Invitrogen™ Supplier No. D16

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

5-(4,6-dichlorotriazinyl)aminofluorescein (5-DTAF) is a reactive dye with absorption/emission maxima of ∼492/516 nm. Dichlorotriazines readily modify amines in proteins, and are among the few reactive groups that are reported to react directly with polysaccharides and other alcohols in aqueous solution, provided that the pH is >9 and that other nucleophiles are absent.

5-(4,6-dichlorotriazinyl)aminofluorescein (5-DTAF) is a reactive dye with absorption/emission maxima of ∼492/516 nm. Dichlorotriazines readily modify amines in proteins, and are among the few reactive groups that are reported to react directly with polysaccharides and other alcohols in aqueous solution, provided that the pH is >9 and that other nucleophiles are absent.

Specifications

Quantity 100 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Alcohol, Amine
Shipping Condition Room Temperature
Reactive Moiety Dichlorotriazine
Label or Dye FITC (Fluorescein)
Label Type Classic Dyes

For Research Use Only. Not for use in diagnostic procedures.