missing translation for 'onlineSavingsMsg'
Learn More

Span 85 (=Sorbitan Trioleate), TCI America™

$41.02 - $106.25

Chemical Identifiers

CAS 26266-58-0
MDL Number MFCD00133820
Synonym Arlacel 85, Sorbitan Trioleate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S006425G
View Documents
TCI America
S006425G
25 g
Each for $41.02
Only null left
 
S0064500G
View Documents
TCI America
S0064500G
500 g
Each for $106.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

26266-58-0
MFCD00133820
Arlacel 85, Sorbitan Trioleate

Specifications

26266-58-0
MFCD00133820
Arlacel 85, Sorbitan Trioleate
Span 85 (=Sorbitan Trioleate)
-23°C
25 g
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (8)-0063

RTECSNumber : WG2934550

TSCA : Yes

SDS
Documents

Documents