missing translation for 'onlineSavingsMsg'
Learn More

Span 85 (=Sorbitan Trioleate), TCI America™

Catalog No. S006425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S006425G 25 g
S0064500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S006425G Supplier TCI America Supplier No. S006425G
Only null left

Chemical Identifiers

CAS 26266-58-0
MDL Number MFCD00133820
Synonym Arlacel 85, Sorbitan Trioleate

Specifications

Melting Point -23°C
Quantity 25 g
Physical Form Liquid
Chemical Name or Material Span 85 (=Sorbitan Trioleate)
EINECSNumber (8)-0063
RTECSNumber WG2934550
TSCA Yes