missing translation for 'onlineSavingsMsg'
Learn More

Span™ 80, Viscosity, 1000-2000 mPa. (20°C), MilliporeSigma™ Supelco™

Catalog No. 111012964
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012964 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012964 Supplier Merck Emd Millipore Supplier No. 855481L
Only null left

Span(R) 80 is a sorbitan ester series of surfactant. It is produced by esterification of one or more sorbitan hydroxyl group with fatty acid. It is hydrophobic in nature.

Specifications

Product Line Span
CAS 1338-43-8
Density 0.99 g/mL at 20°C; 0.986 g/mL (at 25°C)
Flash Point >113°C
Linear Formula C24H44O6
MDL Number MFCD00080948
Synonym Sorbitan monooleate; Span(R) 80
Type Viscosity Standard
Quantity 1 L
Formula Weight 428.60

Span is a registered trademark of Croda International PLC