missing translation for 'onlineSavingsMsg'
Learn More

Span™ 80, Viscosity 1000-2,000 mPa's (20°C), MilliporeSigma™ Supelco™

Catalog No. 111012963
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012963 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012963 Supplier Merck Emd Millipore Supplier No. 85548250ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Span™ 80 is a sorbitan ester series of surfactant. It is produced by esterification of one or more sorbitan hydroxyl group with fatty acid. It is hydrophobic in nature.

Specifications

CAS 1338-43-8
Linear Formula C24H44O6
MDL Number MFCD00080948
Density 0.986 g/mL (at 25°C)
Synonym Sorbitan monooleate; Span(R) 80
Formula Weight 428.60
Refractive Index n20/D 1.48
Quantity 250 mL

Span is a registered trademark of Croda International PLC