missing translation for 'onlineSavingsMsg'
Learn More

SPADNS

CAS: 23647-14-5 | C16H10N2Na3O11S3+ | 571.408 g/mol

Supplier:  Thermo Scientific Chemicals A1693318

 View more versions of this product

Catalog No. AAA1693318


Only null left

Description

Description

Sulfanilic acid azochromotrop is used as an indicator for complexometric determinations of zirconium and thorium. It is used as a reagent for drinking and wastewater analysis using Hach Method 8029, Fluoride, SPADNS Method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfanilic acid azochromotrop is used as an indicator for complexometric determinations of zirconium and thorium. It is used as a reagent for drinking and wastewater analysis using Hach Method 8029, Fluoride, SPADNS Method.

Solubility
Soluble in water (10 mg/ml), and ethanol (1 mg/ml). very slightly soluble or insoluble in alcohol and organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
Specifications

Specifications

SPADNS
23647-14-5
MFCD00003952
Soluble in water (10mg/ml),and ethanol (1mg/ml). very slightly soluble or insoluble in alcohol and organic solvents.
C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]
571.408
570.41
>300°C
C16H10N2Na3O11S3+
Sulfanilic acid azochromotrope; 2-(4-Sulfophenylazo)chromotropic acid trisodium salt
LSAWBVPBZOEGOT-UHFFFAOYSA-L
trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
73658865
50 g

Chemical Identifiers

23647-14-5
571.408
LSAWBVPBZOEGOT-UHFFFAOYSA-L
73658865
C1=CC(=CC=C1NN=C2C(=CC3=CC(=CC(=C3C2=O)O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O.[Na+].[Na+].[Na+]
C16H10N2Na3O11S3+
MFCD00003952
Sulfanilic acid azochromotrope; 2-(4-Sulfophenylazo)chromotropic acid trisodium salt
trisodium;5-hydroxy-4-oxo-3-[(4-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 245-803-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only