missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SP5/SP8/SP9 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SP5/8/9.
Binds to GC boxes promoters elements. Probable transcriptional activator that has a role in the coordination of changes in transcription required to generate pattern in the developing embryo.
Specifications
Specifications
| Antigen | SP5/SP8/SP9 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SP9 |
| Gene Accession No. | P0CG40, Q64HY3, Q6BEB4, Q8BMJ8, Q8IXZ3, Q9JHX2 |
| Gene Alias | BTD; buttonhead-like zinc finger transcription factor Sp9; D930049B17Rik; mBtd; SP5; Sp5 transcription factor; SP8; Sp8 transcription factor; Sp9; Sp9 transcription factor; Sp9 transcription factor homolog; specificity protein 8; trans-acting transcription factor 5; trans-acting transcription factor 8; trans-acting transcription factor 9; transcription factor Sp5; Transcription factor Sp8; transcription factor Sp9; zinc finger protein 990; ZNF990 |
| Gene Symbols | Sp5, Sp8, SP9 |
| Show More |