missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SP Sepharose™ XL

Catalog No. 45002063
Change view
Click to view available options
Quantity:
300 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002063 300 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002063 Supplier Cytiva Supplier No. 17507301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Strong cation exchanger with high loading capacity and chemical stability

  • Bioprocess medium with hydrophilic base matrix
  • Ligand: sulphopropyl
  • Average particle size: 90μm
  • Matrix: 6% cross-linked agarose with dextran surface extenders
  • Ionic capacity: 0.18 to 0.25mmol H+/mL medium
  • pH stability working range: 4 to 13
  • For use in capture steps of biomolecules from clarified feedstocks
  • Available in 300mL bottle

Specifications

Quantity 300 mL
Stability Non-ionic detergents, 1 M NaOH, 6 M Guanidine Hydrochloride
For Use With (Application) For Strong CatIon Exchanger with High Loading Capacity and Chemical Stability
Storage Requirements 4°C to 30°C
Product Type SP Sepharose XL Media
Flow Rate 300 to 500 cm/Hoμr
Particle Size 45 to 165 μm
Ionic Capacity 0.18 to 0.25 mM H+/mL
Includes Certificate of Analysis
Ion Exchange Type Strong CatIon Exchanger
Matrix 6% Cross-Linked Agarose with Dextran surface extenders
pH Range 4 to 13
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphopropyl
Show More Show Less