missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SP Sepharose™ High Performance Media

Catalog No. 45002947
Change view
Click to view available options
Quantity:
75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002947 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002947 Supplier Cytiva Supplier No. 17108701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for high intermediate and polishing steps in downstream processing. Cytiva Life Sciences™ SP Sepharose™ High Performance Media is composed of crosslinked agarose beads.

  • Composed of crosslinked agarose beads with mean diameter of 34μm, modified with sulphopropyl (SP) strong cation exchange groups
  • High resolution, high capacity separation with high recovery
  • Chemical stability for effective CIP/sanitization
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Quantity 75 mL
For Use With (Application) For High Intermediate and Polishing Steps in Downstream Processing
Stability Stable in commonly used Buffers: 8 M Urea, 6 M Guanidine HCl, 70% Ethanol, 1 M NaOH, 1 M Acetic Acid, 30% Isopropanol, 30% Acetonitrile, 2% SDS1
Storage Requirements 4°C to 30°C
Product Type SP Sepharose High Performance Media
Flow Rate 150 cm/Hoμr
Particle Size 24 to 44 μm
Height (English) Bed 3.93 in.
Height (Metric) Bed 10 cm
Ionic Capacity 0.15 to 0.20 mM H+/mL
Ion Exchange Type Strong CatIon Exchanger
Max. Pressure <3 bar
Matrix 6% Cross-Linked Agarose
pH Range 3 to 14
Storage Buffer 20% Ethanol, 0.2 M Sodium Acetate
Ligand Type Sulphopropyl
Show More Show Less