missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Vibrasorb™ Premium Vibration Damping Mounts

Protects electromechanical and optical equipment from exposure to vibration. Premium vibration damping mounts facilitates optimal equipment use.

Supplier:  Bel-Art™ H183860001

 View more versions of this product

Catalog No. 14-676-3

Only null left
Explore available promotions

Description

Description

Protects electromechanical and optical equipment from exposure to vibration. Bel Art™ SP Scienceware™ Vibrasorb™ Premium Vibration Damping Mounts facilitates optimal equipment use.

  • Polished surface resists scratches and chemical attack 6.4cm (2½ in.) thick platforms
  • Molded of dense black & white terrazzo
  • Four Vibro-Absorber™ cushions with neoprene feet support up to 16kg (35 lb)
  • Larger size will absorb vibrations down to 13 Hz with a 16kg (35lb) load; smaller size will absorb vibrations down to 16 Hz. with a 16kg (35lb) load
  • With lighter loads, the minimum frequency of damping for both sizes will increase to a value between 16 Hz. and 25 Hz
  • Shipping weights: large 39kg (87lb), small 19kg (42lb)
Specifications

Specifications

35 x 35 x 7.6 cm
35 cm
Gray
Vibration Damping Mount
Not Autoclavable
Terrazo
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions