missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Two-Color Wash Bottles, Safety-Vented™ and Safety Labeled, Wide-Mouth
Unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know safety-labeled wash bottles.
$106.23 - $124.07
Specifications
| Sterility | Non-sterile |
|---|---|
| Closure Included | Yes |
| Disposable | Reusable |
| Graduated | No |
| Mouth | Wide |
Alert:
Use only with solvents compatible with LDPE.
| Catalog Number | Mfr. No. | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Chemical Label | Price | Quantity & Availability | ||||
|
1139156
|
Bel-Art™
F116430238 |
250 mL | Custom Write-on |
Pack of 3 Each for $106.23
|
|
||||
|
011890106
|
Bel-Art™
F116420638 |
500 mL | Custom Write-on |
Pack of 3 Each for $124.07
|
|
||||
|
011890104
|
Bel-Art™
F116420624 |
500 mL | Isopropanol |
Pack of 3 Each for $124.07
|
|
||||
Description
A unique, 2-piece valve prevents pressure build-up of solvents within these Right-to-Know Safety-Labeled Wash Bottles
- Pre-printed with chemical name and formula, the National Fire Protection Association (NFPA) section 704 four-color diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
- Valve eliminates drips, allows dispensing of wash bottle in an upright or inverted position.
- Wide mouth 53 mm (2 in.) bottle for easy refilling.
- Polypropylene closure with leakproof design, color-coded to aid in easy identification.
- Translucent, LDPE solvent-resistant Sodium Hypochlorite is white LDPE for light-protection between 230 and 450.
- Toluene is red LDPE bottle.
- Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200.
- Labeled with six of the most commonly used solvent names or custom labeling available.
Order Info
8 oz. (250 mL)
Specifications
| Non-sterile | |
| Reusable | |
| Wide | |
| Polypropylene |
| Yes | |
| No | |
| Cylindrical | |
| Not Autoclavable |