Learn More
Bel Art™ Space-Saver Desiccator
For dry storage and protection of moisture-sensitive products.
$95.45 - $625.65
Specifications
Cabinet Style Top Loading
Material Polypropylene, Polycarbonate
Product Type Desiccator
View More Specs
Products 3
Catalog Number Mfr. No. Volume (English) Max. Clearance Above Plate Diameter (Metric) Inner Price Quantity & Availability  
Catalog Number Mfr. No. Volume (English) Max. Clearance Above Plate Diameter (Metric) Inner Price Quantity & Availability  
10987059
View Documents Promotion Details
Bel Art™
F420310000
0.20 cu. ft. 15.7 cm 19.7 cm
Each for $246.46
Case of 4 Each for $985.84
Only null left
16800552
View Documents Promotion Details
Bel Art™
F420300000
0.09 cu. ft. 12.1 cm 14.9 cm
Each for $95.45
Case of 4 Each for $381.80
Only null left
22261909
View Documents Promotion Details
Bel Art™
F420320000
0.31 cu. ft. 19.7 cm 24 cm
Case of 4 Each for $625.65
Only null left
Description
Description
  • Strong and shatterproof with extra heavy walls
  • Flat dome, clear polycarbonate top maximizes interior clearances and provides greater interior volume
  • Large ribbed knob for easy lifting of top
  • Includes 0.32cm (0.125 in.) thick perforated plate
  • Disposable or reusable desiccant can be placed on or under perforated plate
  • Neoprene O-ring ensures tight seal to 0°C without need for grease
  • White polypropylene bottom supports incandescent crucibles on High Heat Minerit HD Ceramic Desiccator Plates
Specifications
Specifications
Top Loading
Polypropylene, Polycarbonate
Desiccator