missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ HDPE Indicator Bottles

Double-duty bottles offer two ways to assist in staining slides.

Specifications

Color Translucent, White
For Use With (Application) Indicator solutions or dyes
Material Polyethylene
No. per Pack 12
Sterility Non-sterile
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S31707
View Documents Promotion Details
Bel-Art™
F11661
30 mL N/A
Only null left
 
S31708
View Documents Promotion Details
Bel-Art™
S31708
60 mL N/A
Only null left
 
Description

Description

  • Can be used as squirt bottles with the supplied tube and as dropper bottles when the tube is removed
  • Ideal for use with indicator solutions or dyes
Specifications

Specifications

Translucent, White
Polyethylene
Non-sterile
Reusable
No
Cylindrical
Polypropylene
Not Autoclavable
Indicator solutions or dyes
12
Yes
0.002 oz.
Narrow, Threaded
Dispensing Bottle
Threaded
Videos
SDS
Documents

Documents