missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Goat anti-Mouse, Human ads, HRP, Polyclonal, Southern Biotech™
Description
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
Specifications
| Antigen | IgG |
| Applications | ELISA, Dot Blot, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | Lot specific |
| Conjugate | HRP |
| Formulation | Supplied in a stock solution in 50% glycerol/50% PBS, pH 7.4; No preservative added |
| Host Species | Goat |
| Immunogen | Mouse IgG |
| Purification Method | Affinity Purified |
| Quantity | 1.0 mL |
| Show More |