missing translation for 'onlineSavingsMsg'
Learn More

Sotalol hydrochloride, 98%

Catalog No. AAJ63772MC
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63772MC 100 mg
AAJ6377203 1 g
AAJ6377206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ63772MC Supplier Thermo Scientific Chemicals Supplier No. J63772MC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potent beta adrenergic antagonist | CAS: 959-24-0 | C12H21ClN2O3S | 308.82 g/mol

Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.

Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 959-24-0
Molecular Formula C12H21ClN2O3S
Molecular Weight (g/mol) 308.82
MDL Number MFCD00242937
InChI Key VIDRYROWYFWGSY-UHFFFAOYNA-N
Synonym sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
PubChem CID 66245
ChEBI CHEBI:9207
IUPAC Name hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

Specifications

Color White
Quantity 100 mg
Merck Index 14,8728
Solubility Information Soluble in phosphate buffered saline,DMSO,ethanol,water,and methanol.
Formula Weight 308.82
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Sotalol hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 213-496-0
RTECSNumber PB0826000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only