Learn More
Thermo Scientific™ Sorvall™ Legend™ T/RT Centrifuge Buckets for 4-Place Swinging Bucket Rotors
Round or rectangular buckets for use with Four-Place Swinging Bucket Rotors 75006445 and 75006434
$2,104.44 - $2,271.27
Description
Order Info
Sold individually; order correct quantity to equip rotor. Sealing lids sold separately. Accessory adapters accept assorted tube styles and volumes from 1.5 to 250mL.
Specifications
| Swinging bucket rotor | |
| Bucket |